Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Rotary Evaporator Anti-Splash Adapters
GREENER_CHOICESDP

Catalog No. 31500621 Shop All Kemtech Products
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
Standard Taper Joint:
Upper: 24/40, Lower: 14/20
Upper: 24/40, Lower: 19/22
Upper: 24/40, Lower: 24/40
Upper: 29/42, Lower: 14/20
Upper: 29/42, Lower: 24/40
Upper: 29/42, Lower: 29/42
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Standard Taper Joint
31-500-621 250 mL Upper: 29/42, Lower: 29/42
31-500-612 100 mL Upper: 24/40, Lower: 14/20
31-500-613 100 mL Upper: 24/40, Lower: 19/22
31-500-614 100 mL Upper: 24/40, Lower: 24/40
31-500-615 100 mL Upper: 29/42, Lower: 14/20
31-500-616 100 mL Upper: 29/42, Lower: 24/40
31-500-617 250 mL Upper: 24/40, Lower: 14/20
31-500-618 250 mL Upper: 24/40, Lower: 19/22
31-500-619 250 mL Upper: 24/40, Lower: 24/40
31-500-620 250 mL Upper: 29/42, Lower: 24/40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 31-500-621 Supplier Synthware™ Supplier No. A288250
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Prevents possible contamination from rotary evaporator

  • Placed between vapor tube and evaporation flask on rotary evoprator
  • Prevent contents of flask to be drawn into condensor in event of bumping

Specifications

Standard Taper Joint Upper: 29/42, Lower: 29/42
Product Type Anti-splash Adapter
Material Borosilicate Glass (3.3)
Capacity (Metric) 250 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.