Learn More
Description
- SYNTHWARE round bottom boiling flasks with full length standard taper outer joint
- All sizes are heavy wall design and are hand blown from borosilicate tubing to ensure uniform wall thickness
Specifications
Specifications
| Color | Clear |
| Description | Single neck, round bottom flask |
| Capacity (Metric) | 250 mL |
| Material | Borosilicate Glass (3.3) |
| For Use With (Application) | Organic Synthesis |
| For Use With (Equipment) | Organic Chemistry 14/20 Kit |
| Product Type | Boiling Flask |
| Standard Taper Joint | 14/20 |
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

