Learn More
Synthware™ Thermometer Adapter
GREENER_CHOICESDP
Catalog No. 31500578 Shop All Kemtech Products
Change view
Click to view available options
Quantity:
1
Catalog No. Quantity
31-500-578 1
1 options
Catalog No. 31-500-578 Supplier Synthware™ Supplier No. A162440
Add to Cart
Add to Cart
With 10/30 thermometer joint, angled for use in multiple neck flask
Specifications
For Use With (Application) Organic Synthesis
Material Borosilicate Glass (3.3)
Product Type Thermometer adapter
Quantity 1
Standard Taper Joint No. 24/40