Learn More
Synthware™ U-Shape Adapter
GREENER_CHOICESDP
Catalog No. 31500699 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint No.:
14/20
Catalog No. Standard Taper Joint No.
31-500-699 14/20
1 options
Catalog No. 31-500-699 Supplier Synthware™ Supplier No. A631420
Add to Cart
Add to Cart
With standard taper inner joints at both ends
Specifications
For Use With (Application) Organic Synthesis
Material Borosilicate Glass (3.3)
Product Type U shape adapter
Quantity 1
Standard Taper Joint No. 14/20