Learn More
Synthware™ Vacuum Adapters with Outer Joint
GREENER_CHOICESDP
Catalog No. 31500553 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint No.:
14/20
24/40
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No.
31-500-553 24/40
31-500-552 14/20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 31-500-553 Supplier Synthware™ Supplier No. A322440
Add to Cart
Add to Cart
Ideal for use as a vent or for the introduction of pressure or vacuum
  • Synthware vacuum adapter has a standard taper outer joint and a serrated hose connection
  • Ideal for use as a vent or for the introduction of pressure/vacuum
Specifications
Description Vacuum Adapter w/ Outer Joint
For Use With (Application) Organic Synthesis
Fits Tubing ID (Metric) Hose O.D.: 10 mm
Material Borosilicate Glass (3.3)
Product Type Vacuum Adapter with Outer Joint
Quantity 1
Standard Taper Joint No. 24/40