Learn More
Synthware™ Vacuum or Argon Adapters
GREENER_CHOICESDP
Catalog No. 31500531 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint No.:
14/20
19/22
24/40
29/42
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No.
31-500-531 14/20
31-500-532 19/22
31-500-533 24/40
31-500-534 29/42
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 31-500-531 Supplier Synthware™ Supplier No. A291420
Add to Cart
Edge
Add to Cart
With hose connection on the side and male joint
  • Synthware vacuum or inert gas adapter.
  • Allows for flexible tubing to be connected to apparatus for introduction of vacuum or pressure.
  • Hose connection is at a 90 degree angle to the standard taper inner joint.
Specifications
For Use With (Application) Organic Synthesis
Material Borosilicate Glass (3.3)
Product Type Vacuum or Argon Adapter
Quantity 1
Standard Taper Joint No. 14/20