Learn More
Synthware™ Vacuum or Gas Adapters
GREENER_CHOICESDP
Catalog No. 31500538 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint No.:
14/20
19/22
24/40
29/42
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No.
31-500-538 24/40
31-500-536 14/20
31-500-537 19/22
31-500-539 29/42
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 31-500-538 Supplier Synthware™ Supplier No. A302440
Add to Cart
Add to Cart
With hose connection on the top and male joint
Synthware vacuum or argon/nitrogen inlet adapter. Allows for flexible tubing to be connected to apparatus for introduction of vacuum or pressure.
Specifications
For Use With (Application) Organic Synthesis
Material Borosilicate Glass (3.3)
Product Type Vacuum or Gas Adapter
Quantity 1
Standard Taper Joint No. 24/40