Description
- Synthware vacuum or gas inlet adapter
- Allows for flexible tubing to be connected to apparatus for introduction of vacuum or pressure
- Hose connection is at a 90 degree angle to the standard taper outer joint
Specifications
Specifications
| Description | Vacuum/Inert Gas Adapter, 90 Degree, with Outer Joint |
| For Use With (Application) | Organic Synthesis |
| Fits Tubing ID (Metric) | Hose O.D.: 8 mm |
| Material | Borosilicate Glass (3.3) |
| Product Type | Vacuum/Inert Gas Adapter, 90 Degree, with Outer Joint |
| Quantity | 1 |
| Standard Taper Joint No. | 19/22 |