Description
- 10/18 top joint accommodates 76mm immersion thermometer
Specifications
Specifications
| Material | Borosilicate Glass (3.3) |
| Product Type | Distillation head adapter |
| Standard Taper Joint No. | 10/18 Top, 24/40 Other |