Description
- Synthware vacuum transfer adapter with PTFE stopcocks and standard taper inner joint
- Used for the transfer of sensitive and hazardous solutions while under vacuum
Specifications
Specifications
| For Use With (Application) | Organic Synthesis |
| Material | Borosilicate Glass (3.3) |
| Product Type | Glass Joint Adapter |
| Quantity | 1 |
| Standard Taper Joint No. | 24/40 |