Learn More
Sigma Aldrich Fine Chemicals Biosciences SynthwareTM all glass vacu1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences SYNM1810051EA
Encompass
With 4 mm double oblique high vacuum hollow glass stopcocks precision ground with corresponding numbers on barrel and plug. Lower vacuum chambers on stopcock are evacuated with a separate hole in the plug in order to hold the plug in the stopcock barrels. Beams are 22 mmO.D. with front-left and rear-right 10 mm hose connections. Distance between stopcocks is 75 mm center to center.
Catalog No. 50-290-8681