Learn More
Sigma Aldrich Fine Chemicals Biosciences SynthwareTM all glass vacu1EA
Catalog No. 502908738
Encompass
With 4 mm double oblique high vacuum solid glass stopcocks precision ground with corresponding numbers on barrel and plug. Solid glass stopcocks have constant pressure retention devices. Beams are 22 mm O.D. with front-left-right and rear-left 10 mm hose connections. Distance between stopcocks is 75 mm center to center.
Catalog No. 50-290-8738 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SYNM2600051EA
With 4 mm double oblique high vacuum solid glass stopcocks precision ground with corresponding numbers on barrel and plug. Solid glass stopcocks have constant pressure retention devices. Beams are 22 mm O.D. with front-left-right and rear-left 10 mm hose connections. Distance between stopcocks is 75 mm center to center.