Learn More
Sigma Aldrich Fine Chemicals Biosciences SynthwareTM gas washing bo1EA
Catalog No. 502906595
Encompass
Gas washing bottle with standard taper 45/50 cap style stopper and fritted disc angled at 15 for even distribution of gas throughout absorbing material. Available with a coarse or extra coarse fritted disc. Order plastic clip separately. (not supplied)
Catalog No. 50-290-6595 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SYNB300250C1EA
Gas washing bottle with standard taper 45/50 cap style stopper and fritted disc angled at 15 for even distribution of gas throughout absorbing material. Available with a coarse or extra coarse fritted disc. Order plastic clip separately. (not supplied)