Learn More
Sigma Aldrich Fine Chemicals Biosciences SynthwareTM single-neck ro1EA
Catalog No. 502900206
Encompass
Heavy wall design. All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
Catalog No. 50-290-0206 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SYNF3045001EA
Add to Cart
Add to Cart
Heavy wall design. All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.