Learn More
Sigma Aldrich Fine Chemicals Biosciences SynthwareTM Vacuum/Inert G1EA
Catalog No. 502908873
Encompass
General purpose double bank manifold for use with vacuum and inert gas. Supplied with 0-4 mm high vacuum valves for grease-free operation. Both banks are made from 22 mm medium wall tubing and have all banks open. Ports are spaced 100 mm center to center and have an O.D. of 10 mm. With front- right and rear- left 10 mm hose connections.
Catalog No. 50-290-8873 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SYNM1410041EA
General purpose double bank manifold for use with vacuum and inert gas. Supplied with 0-4 mm high vacuum valves for grease-free operation. Both banks are made from 22 mm medium wall tubing and have all banks open. Ports are spaced 100 mm center to center and have an O.D. of 10 mm. With front- right and rear- left 10 mm hose connections.