Learn More
CPC Scientific H-Pro-Leu-Ala-Arg-Thr-Leu-Ser-Val-Ala-Gly-Leu-Pro-Gly-Lys-Lys-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific PKCP050A
Encompass
ONE-LETTER SEQUENCE: PLARTLSVAGLPGKKMOLECULAR FORMULA: C68H122N20O18MOLECULAR WEIGHT:1507.84STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [108334-68-5], SYNONYMS: Calmodulin-Dependent Protein Kinase SubstrateRESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES: Y. Hashimoto et al., Arch. Biochem. Biophys., 252, 418 (1987)
Catalog No. 50-211-0205