Learn More
Medchemexpress LLC HY-124876 5mg Medchemexpress, SynuClean-D CAS:685121-45-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1248765MG
Encompass_Preferred
Medchemexpress, HY-124876 5mg SynuClean-D CAS:685121-45-3 SynuClean-D (SC-D) is an inhibitor of α-synuclein aggregation, disrupts mature amyloid fibrils, prevents fibril propagation, and abolishes the degeneration of dopaminergic neurons in an animal model of Parkinson’s disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0013