Learn More
Medchemexpress LLC α-Synuclein (61-75) TFA | 99.3% | 1544.58 | 25 MG
SDP
Catalog No. 5000397416
Encompass_Preferred
α-Synuclein (61-75) TFA is a fragment of α-Synuclein, an abundant neuronal protein highly enriched in presynaptic nerve terminals. It is identified as a potential biomarker for Parkinson's disease (PD).
  • Fragment of α-synuclein
  • Abundant neuronal protein
  • Highly enriched in presynaptic nerve terminals
  • Potential biomarker for Parkinson's disease
Catalog No. 50-003-97416 Supplier Medchemexpress LLC Supplier No. 25MGHYP3140A
α-Synuclein (61-75) TFA is a fragment of α-Synuclein, an abundant neuronal protein highly enriched in presynaptic nerve terminals. It is identified as a potential biomarker for Parkinson's disease (PD).
  • Fragment of α-synuclein
  • Abundant neuronal protein
  • Highly enriched in presynaptic nerve terminals
  • Potential biomarker for Parkinson's disease