Learn More
Medchemexpress LLC Α-Synuclein (61-95) (human) | 154040-19-4 | 3260.61 | 500 UG
SDP
Catalog No. 5000396532
Encompass_Preferred
α-Synuclein (61-95) (human) is the hydrophobic core region of α-synuclein that induces neuronal cell death. This product is for research use only and can be utilized in studies of neurodegenerative diseases.
  • Induces neuronal cell death
  • Suitable for research of neurodegenerative diseases
  • Applicable to Alzheimer's disease (AD) research
  • Applicable to Parkinson's disease (PD) research
Catalog No. 50-003-96532 Supplier Medchemexpress LLC Supplier No. HYP4704A500UG
α-Synuclein (61-95) (human) is the hydrophobic core region of α-synuclein that induces neuronal cell death. This product is for research use only and can be utilized in studies of neurodegenerative diseases.
  • Induces neuronal cell death
  • Suitable for research of neurodegenerative diseases
  • Applicable to Alzheimer's disease (AD) research
  • Applicable to Parkinson's disease (PD) research