Learn More
Aviva Systems Biology Corp SYPL1 Antibody - C-terminal region (ARP46774_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46774_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SYPL1. It was validated on Western Blot. . Alternate names: H-SP1, SYPLApplicationWB. Species reactivity: Dog, Horse, Human, Pig
Catalog No. 50-285-273