Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ SYPRO™ Protein Gel Stains

Catalog No. BMA50562 Shop All Lonza, Inc Products
Change view
Click to view available options
Color:
Ruby
Quantity:
200 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Quantity
BMA50562 Ruby 1 L
BMA50564 Ruby 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BMA50562 Supplier Lonza™ Supplier No. 50562
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Liquid format protein gel stains

SYPRO Ruby Protein Gel Stains

  • Ideal for 2-D gel analysis Sensitivity rivals silver stain Detects nanogram levels of protein Offers a broad linear quantitation range and consistent gel-to-gel staining for accurate protein expression comparisons
SYPRO Tangerine Protein Gel Stain
  • Provides high sensitivity for detection of proteins in SDS-polyacrylamide gels
  • Does not require the use of acids or organic solvents
  • Suitable for use with saline solutions Ideal for use when eluting active protein complexes from gels because the mild staining conditions are unlikely to affect protein activity
SYPRO Red Protein Gel Stain
  • Detects as little as 4 to 8ng of protein per band or proteins as small as 6.5kDa Provides linear protein quantitation up to three orders of magnitude, from ng to μg per band
  • Offers complete staining in less than one hour—no fixation or destaining required
  • Compatible with many types of detection systems Recommended for instruments that employ frequency-doubled Nd:YAG or green or red He-Ne lasers

Product licensed by Cambrex from Molecular Probes, Inc. For research and quality control use only.

TRUSTED_SUSTAINABILITY

Specifications

Color Ruby
Form Liquid
Quantity 1 L
Storage Requirements 4°C
Product Type Protein Stain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.