Learn More
Med Vet International AHS Syringe & Needle, .5mL, Permanent Needle, 29G X 1/2in, U-100 Insulin, 1000/CS, AH05M2913
Catalog No. 501180853
Encompass_Preferred
AHS AH05M2913 .5mL Syringe & Needle Combo comes with a clear barrel and easy to read graduation marks. This syringe contains a permanently attached needle. This AHS Needle is a 29G X 1/2' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This is an insulin syringe. It is used to inject insulin in the subcutaneous fat. This quantity is 1000/CS
Catalog No. 50-118-0853 Supplier Med Vet International Supplier No. AH05M2913X10
AHS AH05M2913 .5mL Syringe & Needle Combo comes with a clear barrel and easy to read graduation marks. This syringe contains a permanently attached needle. This AHS Needle is a 29G X 1/2' Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This is an insulin syringe. It is used to inject insulin in the subcutaneous fat. This quantity is 1000/CS