Learn More
Trajan Scientific SYR 100UL 100R-C/F-GT-LC
SDP
Catalog No. 502530328
Encompass_Preferred
Trajan Autosampler Syringes, Connection Type: Removable, Volume: 100 uL, Needle Gauge: 22 G, For Use With: Carlo Erba/Fisons/Leap/CTC ASD200 and AS800, High performance liquid chromatology, Length Needle: 51 mm, 2 in., Diameter Outer Needle: 0.02 in., 0.72 mm, Model: 100R-C/T-GT-LC
Catalog No. 50-253-0328 Supplier Trajan Scientific Supplier No. 005330
Trajan Autosampler Syringes, Connection Type: Removable, Volume: 100 uL, Needle Gauge: 22 G, For Use With: Carlo Erba/Fisons/Leap/CTC ASD200 and AS800, High performance liquid chromatology, Length Needle: 51 mm, 2 in., Diameter Outer Needle: 0.02 in., 0.72 mm, Model: 100R-C/T-GT-LC