Learn More
Trajan Scientific SYR 100UL LEUR TIP
SDP
Catalog No. 502530333
Encompass_Preferred
Trajan Fixed Luer Tip Syringes, Volume: 100 uL, Connection Type: Fixed, For Use With: For maximum inertness, Borosilicate Glass, Termination: Luer-tip, Manual Syringe
Catalog No. 50-253-0333 Supplier Trajan Scientific Supplier No. 005229
Trajan Fixed Luer Tip Syringes, Volume: 100 uL, Connection Type: Fixed, For Use With: For maximum inertness, Borosilicate Glass, Termination: Luer-tip, Manual Syringe