Learn More
Trajan Scientific SYR 10MDR-HSV
SDP
Catalog No. 502530294
Encompass_Preferred
SGE Gas Tight Plunger Syringe with PTFE Luer Lock, Volume: 10.0 mL, For Use With: For soil gas sampling, Includes: PTFE luer-lock valve, Quantity: Each
Catalog No. 50-253-0294 Supplier Trajan Scientific Supplier No. 008975
SGE Gas Tight Plunger Syringe with PTFE Luer Lock, Volume: 10.0 mL, For Use With: For soil gas sampling, Includes: PTFE luer-lock valve, Quantity: Each