Learn More
Trajan Scientific SYR 10ML 10MDF-LL-GT
SDP
Catalog No. 502530296
Encompass_Preferred
SGE Gastight Syringes: Fixed Luer-Lok Models, Volume: 10.0 mL, Diameter Outer: 0.7 in., 18 mm, Model: 10MDF-LL-GT, Accuracy: +/-1%, Connection Type: Fixed, For Use With: Suitable for gases or liquids for maximum inertness, Includes: Plunger protection, inert Kel-F male luer cone
Catalog No. 50-253-0296 Supplier Trajan Scientific Supplier No. 008962
SGE Gastight Syringes: Fixed Luer-Lok Models, Volume: 10.0 mL, Diameter Outer: 0.7 in., 18 mm, Model: 10MDF-LL-GT, Accuracy: +/-1%, Connection Type: Fixed, For Use With: Suitable for gases or liquids for maximum inertness, Includes: Plunger protection, inert Kel-F male luer cone