Learn More
Trajan Scientific SYR 10R-S-GT-0.63
SDP
Catalog No. 502530371
Encompass_Preferred
SGE Gas Tight Syringe with Removable Needle, Length: 1.65 in., 42 mm, Length Needle: 1.65 in., 42 mm, Model: 10R-S-GT-0.63C, Needle Point Style: Removable Cone, Needle Gauge: 23 G, Volume: 10 uL, Connection Type: Removable, Diameter Outer Needle: 0.024 in., 0.63 mm
Catalog No. 50-253-0371 Supplier Trajan Scientific Supplier No. 002902
SGE Gas Tight Syringe with Removable Needle, Length: 1.65 in., 42 mm, Length Needle: 1.65 in., 42 mm, Model: 10R-S-GT-0.63C, Needle Point Style: Removable Cone, Needle Gauge: 23 G, Volume: 10 uL, Connection Type: Removable, Diameter Outer Needle: 0.024 in., 0.63 mm