Learn More
Trajan Scientific SYR 10UL 10F-C/F-GT-5/0.63
SDP
Catalog No. 502530355
Encompass_Preferred
SGE PAL Autosampler Syringes with 50mm Fixed Cone Tip Needle, Description: Gas tight, Volume: 10 uL, Diameter Outer Needle: 0.63 mm, Needle Gauge: 23 G, Length Needle: 50 mm, Connection Type: Fixed, For Use With: Gas chromatography, CTC, LEAP, CombiPAL, GC PAL, Thermo TriPlus & AS2000, Length: 50 mm
Catalog No. 50-253-0355 Supplier Trajan Scientific Supplier No. 002987
SGE PAL Autosampler Syringes with 50mm Fixed Cone Tip Needle, Description: Gas tight, Volume: 10 uL, Diameter Outer Needle: 0.63 mm, Needle Gauge: 23 G, Length Needle: 50 mm, Connection Type: Fixed, For Use With: Gas chromatography, CTC, LEAP, CombiPAL, GC PAL, Thermo TriPlus & AS2000, Length: 50 mm