Learn More
Neta Scientific SYR 10UL FN PTFE TIP 2342HP
SDP
Catalog No. 50929109
Encompass_Preferred
This item is manufactured by Agilent
Catalog No. 50-929-109 Supplier Neta Scientific Supplier No. 51818809
This item is manufactured by Agilent