Learn More
Trajan Scientific SYR 10UL GT HP 23/26 6/PK
SDP
Supplier:  Trajan Scientific 002827
Encompass_Preferred
Trajan Autosampler Syringes with 42mm Cone Tip Fixed Needle, Volume: 10 uL, Description: Gas tight, Length Needle: 42 mm, Needle Gauge: 23 and 26 G, Connection Type: Fixed, Diameter Inner Needle: 0.11 mm, Diameter Outer: 6.5 mm, Diameter Outer Needle: 0.63/0.47 mm
Catalog No. 50-253-0382