Learn More
Med Vet International AHS Tuberculin Syringe & Needle, 1mL 27G x 1/2in., Luer Slip, 1000/Case, AH01T2713
Catalog No. 503042964
Encompass_Preferred
AHS AH01T2713 1mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Slip tip of this 1mL syringe fits conventional and safety needles. This quantity is 1000/CS
Catalog No. 50-304-2964 Supplier Med Vet International Supplier No. AH01T2713X10
AHS AH01T2713 1mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Slip tip of this 1mL syringe fits conventional and safety needles. This quantity is 1000/CS