Learn More
Med Vet International Nipro Tuberculin Syringe & Needle, 1mL, Luer Slip, 27g X 1/2", 100/Box, Jd+01T2713
Catalog No. 502832146
Encompass_Preferred
Nipro JD+01T2713 1mL Syringe & Needle Combo comes with a clear barrel and easy to read graduation marks. The Luer Slip tip of this 1mL syringe fits conventional and safety needles. This Nipro JD+01T2713 Syringe & Needle Combo comes with a 27G X 1/2" Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. Thin wall needles allow rapid and efficient drug delivery. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
Catalog No. 50-283-2146 Supplier Med Vet International Supplier No. JD01T2713
Nipro JD+01T2713 1mL Syringe & Needle Combo comes with a clear barrel and easy to read graduation marks. The Luer Slip tip of this 1mL syringe fits conventional and safety needles. This Nipro JD+01T2713 Syringe & Needle Combo comes with a 27G X 1/2" Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. Thin wall needles allow rapid and efficient drug delivery. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX