Learn More
Trajan Scientific SYR 2.5ML 2.5MDF-GT
SDP
Catalog No. 502530303
Encompass_Preferred
SGE Gastight Syringes: Fixed Needle Models, Volume: 2.5 mL, Needle Gauge: 22 G, Model: 2.5MDF-GT, Length Needle: 50 mm, 1.96 in., Accuracy: +/-1%, Diameter Outer: 0.42 in., 10.8 mm, Diameter Outer Needle: 0.02 in., 0.72 mm, For Use With: Suitable for gases or liquids for maximum inertness
Catalog No. 50-253-0303 Supplier Trajan Scientific Supplier No. 008502
SGE Gastight Syringes: Fixed Needle Models, Volume: 2.5 mL, Needle Gauge: 22 G, Model: 2.5MDF-GT, Length Needle: 50 mm, 1.96 in., Accuracy: +/-1%, Diameter Outer: 0.42 in., 10.8 mm, Diameter Outer Needle: 0.02 in., 0.72 mm, For Use With: Suitable for gases or liquids for maximum inertness