Learn More
Trajan Scientific SYR 250R-V-GT
SDP
Catalog No. 502530320
Encompass_Preferred
SGE Gas Tight Syringe with Removable Needle, Length: 1.96 in., 50 mm, Length Needle: 1.96 in., 50 mm, Model: 250R-V-GT, Needle Point Style: Fixed Bevel, Needle Gauge: 23 G, Volume: 250 uL, Connection Type: Removable, Diameter Outer Needle: 0.024 in., 0.63 mm
Catalog No. 50-253-0320 Supplier Trajan Scientific Supplier No. 006279
SGE Gas Tight Syringe with Removable Needle, Length: 1.96 in., 50 mm, Length Needle: 1.96 in., 50 mm, Model: 250R-V-GT, Needle Point Style: Fixed Bevel, Needle Gauge: 23 G, Volume: 250 uL, Connection Type: Removable, Diameter Outer Needle: 0.024 in., 0.63 mm