Learn More
Med Vet International AHS Syringe & Needle, 3/10mL, Permanent Needle, 30G X 5/16", U-100 Insulin, 1000/Case, AH03M3013
Catalog No. 503045036
Encompass_Preferred
AHS AH05M2913 3/10mL Syringe & Needle Combo comes with a clear barrel and easy to read graduation marks. This syringe contains a permanently attached needle. This AHS Needle is a 30G X 5/16" Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This is an insulin syringe. It is used to inject insulin in the subcutaneous fat. This quantity is 1000/Case
Catalog No. 50-304-5036 Supplier Med Vet International Supplier No. AH03M3013X10
AHS AH05M2913 3/10mL Syringe & Needle Combo comes with a clear barrel and easy to read graduation marks. This syringe contains a permanently attached needle. This AHS Needle is a 30G X 5/16" Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This is an insulin syringe. It is used to inject insulin in the subcutaneous fat. This quantity is 1000/Case