Learn More
Med Vet International Exel Syringe & Needle, 3mL, Luer Lock, 22G X 3/4", Hypodermic, 100/BX, 26115
Catalog No. 503043433
Encompass_Preferred
Exel International 3mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 3mL syringe fits conventional and safety needles. This Exel International 26115 Syringe & Needle Combo comes with a 22G X 3/4" Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
Catalog No. 50-304-3433 Supplier Med Vet International Supplier No. 26115
Exel International 3mL Syringe comes with a clear barrel and easy to read graduation marks. The Luer Lock tip of this 3mL syringe fits conventional and safety needles. This Exel International 26115 Syringe & Needle Combo comes with a 22G X 3/4" Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX