Learn More
Trajan Scientific SYR 500R-GT-LC SS
SDP
Catalog No. 502530311
Encompass_Preferred
Trajan Syringes for Rheodyne and Valco Valves, Volume: 500 uL, Connection Type: Removable, Length Needle: 51 mm, 2 in., Needle Gauge: 22 G, Accuracy: +/-1%, Diameter Outer Needle: 0.028 in., 0.71 mm, For Use With: For Rheodyne and Valco valves, Length: 2 in., 51 mm
Catalog No. 50-253-0311 Supplier Trajan Scientific Supplier No. 007312
Trajan Syringes for Rheodyne and Valco Valves, Volume: 500 uL, Connection Type: Removable, Length Needle: 51 mm, 2 in., Needle Gauge: 22 G, Accuracy: +/-1%, Diameter Outer Needle: 0.028 in., 0.71 mm, For Use With: For Rheodyne and Valco valves, Length: 2 in., 51 mm