Learn More
Trajan Scientific SYR 50MR-VLLMA-GT
SDP
Supplier:  Trajan Scientific 009670
Encompass_Preferred
SGE Syringes with Valves, Volume: 50 mL, Borosilicate Glass, PTFE Plunger, Autoclavable: Autoclavable, Connection Type: Removable, For Use With: For sample storage and transportation or to pressurize sample prior to injection, Termination: Luer-lock
Catalog No. 50-253-0290