Learn More
Trajan Scientific L-MARK Brand 50l Syringe, Manual, FN, 51mm, For Agilent
Catalog No. 501251203
Encompass_Preferred
L-MARK Brand Syringe, 50-uL Syringe, Manual,Fixed Needle (FN), Point Style. 3, 51mm, 22s, fitted Plunger Similar to Agilent PN 5182-9619 now 5190-1501, 3 Pack Made in Germany.
Catalog No. 50-125-1203 Supplier Trajan Scientific Supplier No. LMK.30301183P
L-MARK Brand Syringe, 50-uL Syringe, Manual,Fixed Needle (FN), Point Style. 3, 51mm, 22s, fitted Plunger Similar to Agilent PN 5182-9619 now 5190-1501, 3 Pack Made in Germany.