Learn More
Trajan Scientific SYR 50UL 50R-HP-4.2/0.63C
SDP
Catalog No. 502530339
Encompass_Preferred
SGE Autosampler Syringes with 42mm Cone Tip Removable Needle, Description: Not gas tight, Volume: 50 uL, Diameter Outer Needle: 0.63 mm, Needle Gauge: 23 G, Length Needle: 42 mm, Connection Type: Removable, For Use With: Gas chromatography, Agilent GC Autosamplers HP7673, 7683, 7693A & 6850ALS
Catalog No. 50-253-0339 Supplier Trajan Scientific Supplier No. 004665
SGE Autosampler Syringes with 42mm Cone Tip Removable Needle, Description: Not gas tight, Volume: 50 uL, Diameter Outer Needle: 0.63 mm, Needle Gauge: 23 G, Length Needle: 42 mm, Connection Type: Removable, For Use With: Gas chromatography, Agilent GC Autosamplers HP7673, 7683, 7693A & 6850ALS