Learn More
Trajan Scientific SYR 5UL 5F-PE-7/0.63C
SDP
Catalog No. 502530413
Encompass_Preferred
Trajan Autosampler Syringes, Connection Type: Fixed, Volume: 5 uL, For Use With: PerkinElmer autosystem, Gas chromatography, Needle Gauge: 23 G, Length Needle: 70 mm, 2.7 in., Diameter Outer Needle: 0.02 in., 0.63 mm, Length: 2.75 in., 70 mm, Model: 5F-PE-GT-0.63C, Needle Point Style: Cone
Catalog No. 50-253-0413 Supplier Trajan Scientific Supplier No. 001954
Trajan Autosampler Syringes, Connection Type: Fixed, Volume: 5 uL, For Use With: PerkinElmer autosystem, Gas chromatography, Needle Gauge: 23 G, Length Needle: 70 mm, 2.7 in., Diameter Outer Needle: 0.02 in., 0.63 mm, Length: 2.75 in., 70 mm, Model: 5F-PE-GT-0.63C, Needle Point Style: Cone