Learn More
Trajan Scientific SYR 5UL 5F-S-4.2/0.63C
SDP
Catalog No. 502530409
Encompass_Preferred
Trajan Autosampler Syringes, Connection Type: Fixed, Volume: 5 uL, For Use With: Shimadzu Models AOC14, AOC17, AOC20, AOC20i, Gas chromatography, Needle Gauge: 23 G, Length Needle: 42 mm, 1.6 in., Diameter Outer Needle: 0.02 in., 0.63 mm, Length: 1.65 in., 42 mm, Model: 5F-S-0.63C
Catalog No. 50-253-0409 Supplier Trajan Scientific Supplier No. 001988
Trajan Autosampler Syringes, Connection Type: Fixed, Volume: 5 uL, For Use With: Shimadzu Models AOC14, AOC17, AOC20, AOC20i, Gas chromatography, Needle Gauge: 23 G, Length Needle: 42 mm, 1.6 in., Diameter Outer Needle: 0.02 in., 0.63 mm, Length: 1.65 in., 42 mm, Model: 5F-S-0.63C