Learn More
Trajan Scientific SYR 5UL HP FIXED 23/26 NDL
SDP
Catalog No. 502530415
Encompass_Preferred
SGE Autosampler Syringes with 42 mm Cone Tip Fixed Needle, 5 uL, Description: Not gas tight, Diameter Outer Needle: 0.63/0.47 mm, Needle Gauge: 23 to 26 G, Connection Type: Fixed, For Use With: Agilent GC Autosamplers HP7673, 7683, 7693A & 6850ALS, Gas chromatography, Needle Point Style: Cone
Catalog No. 50-253-0415 Supplier Trajan Scientific Supplier No. 001821
SGE Autosampler Syringes with 42 mm Cone Tip Fixed Needle, 5 uL, Description: Not gas tight, Diameter Outer Needle: 0.63/0.47 mm, Needle Gauge: 23 to 26 G, Connection Type: Fixed, For Use With: Agilent GC Autosamplers HP7673, 7683, 7693A & 6850ALS, Gas chromatography, Needle Point Style: Cone