Learn More
Med Vet International Allergy Tray PrecisionGlide™ 1 mL 3/8 Inch 27 Gauge Non-Safety Thin Wall, 25/Tray
Catalog No. 503043409
Encompass_Preferred
Manufacturer #    305541 Brand    PrecisionGlide™ Manufacturer    BD Country of Origin    United States Application    Allergy Tray Color Code    Tan Gauge    27 Gauge NDC Number    08290305540 Needle Length    3/8 Inch Needle Material    Stainless Steel Needle Needle Point Style    Regular Bevel Needle Style    Attached Needle Packaging Type    Convenience Tray Safety Feature    NonSafety Sterility    Sterile Syringe Material    Plastic Syringe Tip Type    Fixed Needle Tip UNSPSC Code    42183202 Usage    Disposable Volume    1 mL Wall Type    Thin Wall
Catalog No. 50-304-3409 Supplier Med Vet International Supplier No. 305541
Manufacturer #    305541 Brand    PrecisionGlide™ Manufacturer    BD Country of Origin    United States Application    Allergy Tray Color Code    Tan Gauge    27 Gauge NDC Number    08290305540 Needle Length    3/8 Inch Needle Material    Stainless Steel Needle Needle Point Style    Regular Bevel Needle Style    Attached Needle Packaging Type    Convenience Tray Safety Feature    NonSafety Sterility    Sterile Syringe Material    Plastic Syringe Tip Type    Fixed Needle Tip UNSPSC Code    42183202 Usage    Disposable Volume    1 mL Wall Type    Thin Wall