Learn More
Trajan Scientific SYR AUTO SK 5F HP 0.63 6/PK
SDP
Catalog No. 502530416
Encompass_Preferred
SGE Autosampler Syringes with 42 mm Cone Tip Fixed Needle, 5 uL, Description: Not gas tight, Diameter Outer Needle: 0.63 mm, Needle Gauge: 23 G, Connection Type: Fixed, For Use With: Agilent GC Autosamplers HP7673, 7683, 7693A & 6850ALS, Gas chromatography, Needle Point Style: Cone, Volume: 5 uL
Catalog No. 50-253-0416 Supplier Trajan Scientific Supplier No. 001814
SGE Autosampler Syringes with 42 mm Cone Tip Fixed Needle, 5 uL, Description: Not gas tight, Diameter Outer Needle: 0.63 mm, Needle Gauge: 23 G, Connection Type: Fixed, For Use With: Agilent GC Autosamplers HP7673, 7683, 7693A & 6850ALS, Gas chromatography, Needle Point Style: Cone, Volume: 5 uL