Learn More
Med Vet International Enteral / Oral Syringe Vesco 10 mL Enfit Tip Without Safety, VED-610, 500/Case
Catalog No. 503043414
Encompass_Preferred
ISO 80369-3 compliant ENFit connector designed to reduce the risk of tubing misconnections
Catalog No. 50-304-3414 Supplier Med Vet International Supplier No. 1029295CS
ISO 80369-3 compliant ENFit connector designed to reduce the risk of tubing misconnections