Learn More
Tisch Scientific Polypropylene Syringe Filters, 25mm, 10 um, Double Luer Lock, PP Housing, Nonsterile, 100 per pack, SPEC18230
SDP
Supplier:  Tisch Scientific SPEC18230
Encompass
Tisch polypropylene syringe filters a low-cost laboratory solution for well-suited for many needs. They are designed for aqueous or organic solutions that have high levels of debris and for difficult-to-filter solutions. Polypropylene, as a membrane, is ideal for both HPLC chromatography applications and protein recovery analysis applications.
Catalog No. NC2092736