Learn More
Chemglass Life Sciences Syringe Filter, 0.22µm PVDF Membrane, 13mm Housing Diameter, Individually Wrapped, Sterile
SDP
Supplier:  Chemglass Life Sciences CGN2005005
Encompass_Preferred
NEST Syringe Filters are high quality filters individually wrapped and sterile. The standard range of filters are available in PVDF (Polyvinylidene Fluoride) and PES (Polyethersulfone). All filters are designed with Female Luer-Lock inlets and Male Luer Slip outlets. Sterilized non-pyrogenic and RNA/DNA-Free. The syringe filter housings are polypropylene. PVDF ¿ Extremely low protein-binding; for filtration of non-aggressive aqueous and mild organic solutions or were maximizing protein recovery is important
Catalog No. 50-189-7730