Learn More
Chemglass Life Sciences Syringe Filter, 0.45µm PES Membrane, 13mm Housing Diameter, Individually Wrapped, Sterile
SDP
Supplier:  Chemglass Life Sciences CGN2005017
Encompass_Preferred
NEST Syringe Filters are high quality filters individually wrapped and sterile. The standard range of filters are available in PVDF (Polyvinylidene Fluoride) and PES (Polyethersulfone)All filters are designed with Female Luer-Lock inlets and Male Luer Slip outlets. Sterilized non-pyrogenic and RNA/DNA-Free. The syringe filter housings are polypropylene. PES ¿ Low affinity for proteins and extractables with substantially faster flow rates than PVDF; suitable for pre-filtration and filtration of buffers and culture media.
Catalog No. 50-189-7735