Learn More
CELLTREAT Scientific Products CA Syringe Filter, 0.22m with GF Prefilter, 30mm, Sterile
SDP
Supplier:  CELLTREAT Scientific Products 229769
Encompass_Preferred
CELLTREAT Scientific Products 0.22¿m CA w. GF prefilter Syringe Filter. Filters are modled in the polypropylene housings, providing a reliable seal without the use of adhesives. Female luer-lock inlet and make luer slip outlet. Color coded by membrane type for easy identification. CA (Cellulose Acetate). 0.22¿m CA Syringe Filter with integrated Glass Fiber prefilter (229769) is ideal for one-step filtration of viscous solutions and particulate removal. Low protein binding, reduced extractables, and low hold-up volume. Filtration of aqueous solutions such as buffers, media, and reagents. No pre-wetting required. Membrane diameter 30mm. RNAse/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, individually wrapped in peel-to-open paper/plastic blister pack. 30/case.
Catalog No. 50-114-9110